Learn More
MSC Value Collection Divider, Spring Joint, Flat Leg, Steel, Polished, 8 Inch
Catalog No. 5000532551
Encompass
This divider features an 8-inch leg and a spring joint, ideal for accurate measurements. Constructed from steel with a polished finish, it also includes a flat leg with quick nut adjustment for ease of use.
  • Maximum measurement of 8 inch (200 mm)
  • Spring joint type
  • Divider caliper type
  • Flat leg design
  • Steel material
  • Polished finish
  • Quick nut adjustment mechanism
Catalog No. 50-005-32551 Supplier MSC Supplier No. 06485080
This divider features an 8-inch leg and a spring joint, ideal for accurate measurements. Constructed from steel with a polished finish, it also includes a flat leg with quick nut adjustment for ease of use.
  • Maximum measurement of 8 inch (200 mm)
  • Spring joint type
  • Divider caliper type
  • Flat leg design
  • Steel material
  • Polished finish
  • Quick nut adjustment mechanism