Learn More
MSC Value Collection Dock Bumper, Steel, Laminated, 100,000 Lb Capacity
Catalog No. 5000528085
Encompass
This dock bumper is a robust and durable solution designed for docks, featuring a static capacity of 100,000 lbs. It is constructed from steel with a laminated design, ensuring strength and longevity in demanding environments. This dock bumper has a service height range of 39.5 to 51.0 inches.
  • Withstands up to 100,000 Lb of static pressure, providing significant protection for docks and vehicles.
  • Made from steel and features a laminated design for enhanced resilience and extended product life.
  • Offers a service height range of 39.5 to 51.0 inches, suitable for various dock applications.
Catalog No. 50-005-28085 Supplier MSC Supplier No. 47342688
This dock bumper is a robust and durable solution designed for docks, featuring a static capacity of 100,000 lbs. It is constructed from steel with a laminated design, ensuring strength and longevity in demanding environments. This dock bumper has a service height range of 39.5 to 51.0 inches.
  • Withstands up to 100,000 Lb of static pressure, providing significant protection for docks and vehicles.
  • Made from steel and features a laminated design for enhanced resilience and extended product life.
  • Offers a service height range of 39.5 to 51.0 inches, suitable for various dock applications.