Learn More
MSC Value Collection Double Clevis Link, Yellow Chromate, Steel, 1/4 in - 5/16 in
Supplier:  MSC 52683828
Encompass
This Value Collection Double Clevis Link is constructed from yellow chromate steel, designed for reliable performance in various applications. It accommodates chain sizes from 1/4 inch to 5/16 inch and features a substantial load limit of 4,600 Lb. Manufactured in China, this clevis link provides a dependable solution for industrial and commercial requirements.
  • Constructed from durable steel for robust performance.
  • Features a yellow chromate finish for added corrosion resistance.
  • Designed to fit 1/4 in - 5/16 in chain sizes.
  • Offers a high load limit of 4,600 Lb.
  • Manufactured to meet diverse business needs.
Catalog No. 50-005-28152