Learn More
MSC Value Collection Double Loop Chain, 0.0787 in Dia, Zinc Plated, 90 lb
Catalog No. 5000515548
Encompass
This double loop chain from Value Collection is zinc-plated for durability and features a load limit of 90 lb. It has a diameter of 0.0787 in and is suitable for various light-duty applications.
  • 90 lb load limit
  • Zinc plated finish
  • 1.12 in inside length
  • 200 ft overall length
  • Weldless construction
Catalog No. 50-005-15548 Supplier MSC Supplier No. 66795725
This double loop chain from Value Collection is zinc-plated for durability and features a load limit of 90 lb. It has a diameter of 0.0787 in and is suitable for various light-duty applications.
  • 90 lb load limit
  • Zinc plated finish
  • 1.12 in inside length
  • 200 ft overall length
  • Weldless construction