Learn More
MSC Value Collection Drum Cradle, For 15, 30 & 55 gal Drums, 1,000 Lb Capacity
Catalog No. 5000530799
Encompass
This product is a drum cradle designed to hold one drum. It is suitable for 15, 30, and 55-gallon drums and has a weight capacity of 1,000 pounds.
  • Accommodates one drum, specifically 15, 30, and 55-gallon sizes.
  • Can support a load of up to 1,000 pounds.
  • Features a drain height of 20-1/2 inches.
  • Utilizes a manual lift mechanism.
Catalog No. 50-005-30799 Supplier MSC Supplier No. 72756430
This product is a drum cradle designed to hold one drum. It is suitable for 15, 30, and 55-gallon drums and has a weight capacity of 1,000 pounds.
  • Accommodates one drum, specifically 15, 30, and 55-gallon sizes.
  • Can support a load of up to 1,000 pounds.
  • Features a drain height of 20-1/2 inches.
  • Utilizes a manual lift mechanism.