Learn More
MSC Value Collection Drum Pallet Rack, 55 Gal, 1,600 Lb Capacity, For 2 Drums
Catalog No. 5000532595
Encompass
This drum pallet rack provides a robust solution for storing and managing 55 gallon drums. Designed with a 1,600 lb capacity, it can securely hold two drums, optimizing storage space in industrial and laboratory settings.
  • Provides a secure and organized storage solution
  • Features a height of 12-1/2 inch
  • Offers a width of 45-9/32 inch
  • Designed for two 55 gallon drums
  • Supports a 1,600 lb load capacity
Catalog No. 50-005-32595 Supplier MSC Supplier No. 31721863
This drum pallet rack provides a robust solution for storing and managing 55 gallon drums. Designed with a 1,600 lb capacity, it can securely hold two drums, optimizing storage space in industrial and laboratory settings.
  • Provides a secure and organized storage solution
  • Features a height of 12-1/2 inch
  • Offers a width of 45-9/32 inch
  • Designed for two 55 gallon drums
  • Supports a 1,600 lb load capacity