Learn More
MSC Value Collection Duct Elbow, 14 in ID, Galvanized, 1 ea
Catalog No. 5000503868
Encompass
This product consists of 14 in ID, Galvanized Duct Round Adjustable Elbows, with a 26 Gage thickness. It is suitable for ductwork requiring adjustable elbows.
  • Galvanized material for durability
  • Adjustable design for versatile installation
  • Inside diameter of 14 in
  • Thickness of 26 gage
Catalog No. 50-005-03868 Supplier MSC Supplier No. 03378999
This product consists of 14 in ID, Galvanized Duct Round Adjustable Elbows, with a 26 Gage thickness. It is suitable for ductwork requiring adjustable elbows.
  • Galvanized material for durability
  • Adjustable design for versatile installation
  • Inside diameter of 14 in
  • Thickness of 26 gage