Learn More
MSC Value Collection Duct Stack, 2-1/4 in ID, Galvanized, 1 ea
Catalog No. 5000504925
Encompass
This galvanized duct stack is a component designed for use in ventilation and exhaust systems. It features a robust galvanized construction, ensuring durability and efficient air flow within industrial and commercial applications.
  • Inside diameter of 2-1/4 in
  • Constructed from galvanized material
  • Overall length of 12 in
  • Overall height of 60 in
  • Stack type duct fitting
Catalog No. 50-005-04925 Supplier MSC Supplier No. 03379088
This galvanized duct stack is a component designed for use in ventilation and exhaust systems. It features a robust galvanized construction, ensuring durability and efficient air flow within industrial and commercial applications.
  • Inside diameter of 2-1/4 in
  • Constructed from galvanized material
  • Overall length of 12 in
  • Overall height of 60 in
  • Stack type duct fitting