Learn More
MSC Value Collection Electrical Tape, 7 Mil, Rubber Adhesive, 1 ea
Catalog No. 5000788462
Encompass
This electrical tape features a 7 mil thickness and a rubber adhesive, designed for various electrical insulation and bundling applications.
  • Thickness of 7 mil
  • Rubber adhesive
  • Supplied in 20 yd. rolls
Catalog No. 50-007-88462 Supplier MSC Supplier No. 62512165
This electrical tape features a 7 mil thickness and a rubber adhesive, designed for various electrical insulation and bundling applications.
  • Thickness of 7 mil
  • Rubber adhesive
  • Supplied in 20 yd. rolls