Learn More
MSC Value Collection Electronic Caliper Set, 0 to 6 inch, Steel, Digital
Catalog No. 5000536713
Encompass
This electronic caliper set provides precise measurements for various applications. It is a 2-piece set designed for outside diameter measurements, featuring a digital display for accurate readings within a 0 to 6 inch range. Constructed from durable steel, it ensures reliability and longevity in demanding environments.
  • Digital caliper type
  • Measures from 0 to 6 inch
  • Constructed from steel for durability
  • Jaw material made of steel
  • Includes an electronic caliper
  • Uses replacement battery #357
  • Does not include a case
Catalog No. 50-005-36713 Supplier MSC Supplier No. 44310340
This electronic caliper set provides precise measurements for various applications. It is a 2-piece set designed for outside diameter measurements, featuring a digital display for accurate readings within a 0 to 6 inch range. Constructed from durable steel, it ensures reliability and longevity in demanding environments.
  • Digital caliper type
  • Measures from 0 to 6 inch
  • Constructed from steel for durability
  • Jaw material made of steel
  • Includes an electronic caliper
  • Uses replacement battery #357
  • Does not include a case