Learn More
MSC Value Collection Electronic Height Gage, 12 in, 0.0010 in Resolution
Catalog No. 5000514861
Encompass
This Electronic Height Gage is designed for precise measurements. It features a maximum measurement capacity of 12 inches (300.00 mm) and a resolution of 0.0010 inches (0.0100 microns). The device ensures accuracy of 0.040000 mm, with measurements displayed on an LCD screen.
  • Maximum measurement capacity of 12 in (300.00 mm).
  • Resolution of 0.0010 in (0.0100 microns).
  • Accuracy of 0.040000 mm.
  • Features an LCD display type.
  • Operates on an LR44 battery (compatible with SR44 or 357).
  • Generally does not include a magnetic base.
Catalog No. 50-005-14861 Supplier MSC Supplier No. 00449355
This Electronic Height Gage is designed for precise measurements. It features a maximum measurement capacity of 12 inches (300.00 mm) and a resolution of 0.0010 inches (0.0100 microns). The device ensures accuracy of 0.040000 mm, with measurements displayed on an LCD screen.
  • Maximum measurement capacity of 12 in (300.00 mm).
  • Resolution of 0.0010 in (0.0100 microns).
  • Accuracy of 0.040000 mm.
  • Features an LCD display type.
  • Operates on an LR44 battery (compatible with SR44 or 357).
  • Generally does not include a magnetic base.