Learn More
MSC Value Collection Epoxy Wire Shelving Add-On Unit, 24x48x74 in, 4 Shelves
Catalog No. 5000527056
Encompass
This epoxy wire shelving add-on unit is designed for material handling and storage. It features four shelves and specific dimensions, making it suitable for expanding existing shelving systems.
  • 24 in x 48 in x 74 in
  • Add-on unit configuration
  • Four shelves
  • 3200 lb load capacity
  • Epoxy coating
Catalog No. 50-005-27056 Supplier MSC Supplier No. 43381227
This epoxy wire shelving add-on unit is designed for material handling and storage. It features four shelves and specific dimensions, making it suitable for expanding existing shelving systems.
  • 24 in x 48 in x 74 in
  • Add-on unit configuration
  • Four shelves
  • 3200 lb load capacity
  • Epoxy coating