Learn More
MSC Value Collection Feeler Gage Set, 26 Piece, 0.0015 to 0.025 in, Tempered Steel
Catalog No. 5000528482
Encompass
This feeler gage set is designed for precise measurement of gaps and clearances. It features 26 leaves made from tempered steel, offering a wide range of thickness measurements for various applications.
  • Includes 26 individual leaves.
  • Measures thicknesses from 0.0015 to 0.025 in.
  • Each leaf has a length of 3.5 in and a width of 1/2 in.
  • Constructed from durable tempered steel.
  • Features both inch and metric markings.
Catalog No. 50-005-28482 Supplier MSC Supplier No. 06588305
This feeler gage set is designed for precise measurement of gaps and clearances. It features 26 leaves made from tempered steel, offering a wide range of thickness measurements for various applications.
  • Includes 26 individual leaves.
  • Measures thicknesses from 0.0015 to 0.025 in.
  • Each leaf has a length of 3.5 in and a width of 1/2 in.
  • Constructed from durable tempered steel.
  • Features both inch and metric markings.