Learn More
MSC Value Collection Flexible Rope, Polypropylene, 3/16 in, 55 lb Capacity
Catalog No. 5000527067
Encompass
This polypropylene hollow braid flexible rope is designed for various applications requiring a strong yet lightweight rope. It features a hollow braid construction for flexibility and ease of handling.
  • Made from polypropylene material
  • Features a hollow braid construction
  • Measures 3/16 inch in diameter
  • Offers a 55 pound capacity
  • Comes in a yellow color
Catalog No. 50-005-27067 Supplier MSC Supplier No. 45900446
This polypropylene hollow braid flexible rope is designed for various applications requiring a strong yet lightweight rope. It features a hollow braid construction for flexibility and ease of handling.
  • Made from polypropylene material
  • Features a hollow braid construction
  • Measures 3/16 inch in diameter
  • Offers a 55 pound capacity
  • Comes in a yellow color