Learn More
MSC Value Collection Gage Block, Rectangle, 0.1005 in, Grade AS-1, Steel
Catalog No. 5000527609
Encompass
This is a rectangle steel gage block, precisely crafted to 0.1005 " It meets Grade AS-1 accuracy standards and includes NIST certification, ensuring high precision for various measurement applications.
  • Rectangle shape
  • Grade AS-1 accuracy
  • Manufactured from steel
  • Size of 0.1005 inch
  • Includes NIST certification
  • Made in China
Catalog No. 50-005-27609 Supplier MSC Supplier No. 86291051
This is a rectangle steel gage block, precisely crafted to 0.1005 " It meets Grade AS-1 accuracy standards and includes NIST certification, ensuring high precision for various measurement applications.
  • Rectangle shape
  • Grade AS-1 accuracy
  • Manufactured from steel
  • Size of 0.1005 inch
  • Includes NIST certification
  • Made in China