Learn More
MSC Value Collection Gage Block, Rectangle, 0.112 in, Steel, Grade AS-1
Catalog No. 5000515083
Encompass
This rectangular steel gage block is engineered for accurate measurement tasks, boasting an AS-1 accuracy grade. It includes NIST certification, guaranteeing its precision and compliance with national standards.
  • Rectangle shape
  • Manufactured from steel
  • Accuracy grade AS-1
  • NIST certified
Catalog No. 50-005-15083 Supplier MSC Supplier No. 86291655
This rectangular steel gage block is engineered for accurate measurement tasks, boasting an AS-1 accuracy grade. It includes NIST certification, guaranteeing its precision and compliance with national standards.
  • Rectangle shape
  • Manufactured from steel
  • Accuracy grade AS-1
  • NIST certified