Learn More
MSC Value Collection Gage Block, Rectangle, 0.117 inch, Grade 0, Steel
Catalog No. 5000527376
Encompass
This gage block provides precise dimensional measurements for various applications. It is constructed from steel and features a rectangular shape, ensuring reliable performance in demanding environments.
  • Includes NIST certification
  • Rectangle shape
  • Grade 0 accuracy
  • Size is 0.117 inch
  • Made from steel
  • Meets NIST 41111600 standards
Catalog No. 50-005-27376 Supplier MSC Supplier No. 86281904
This gage block provides precise dimensional measurements for various applications. It is constructed from steel and features a rectangular shape, ensuring reliable performance in demanding environments.
  • Includes NIST certification
  • Rectangle shape
  • Grade 0 accuracy
  • Size is 0.117 inch
  • Made from steel
  • Meets NIST 41111600 standards