Learn More
MSC Value Collection Gage Block, Rectangle, 0.148 in, Grade AS-1, Steel
Catalog No. 5000510911
Encompass
This gage block is a precise measuring tool, crafted from durable steel. Designed in a rectangular shape, it offers a size of 0.148 inches with an accuracy grade of AS-1. Each block includes NIST certification, ensuring reliable and traceable standards for calibration and inspection applications.
  • Rectangle shape
  • Made from steel
  • Accuracy grade AS-1
  • Includes NIST certification
Catalog No. 50-005-10911 Supplier MSC Supplier No. 86293404
This gage block is a precise measuring tool, crafted from durable steel. Designed in a rectangular shape, it offers a size of 0.148 inches with an accuracy grade of AS-1. Each block includes NIST certification, ensuring reliable and traceable standards for calibration and inspection applications.
  • Rectangle shape
  • Made from steel
  • Accuracy grade AS-1
  • Includes NIST certification