Learn More
MSC Value Collection Gage Block, Rectangle, 2 in, AS-1, Steel
Catalog No. 5000527624
Encompass
The Value Collection Rectangle Steel Gage Block is a precision measuring tool designed for high accuracy applications. This 2 inch, AS-1 Grade block includes NIST certification, ensuring reliable and traceable measurements for quality control and calibration.
  • Rectangle shape
  • AS-1 accuracy grade
  • 2 inch size
  • Durable steel construction
  • NIST certified for traceability and reliability
Catalog No. 50-005-27624 Supplier MSC Supplier No. 86295805
The Value Collection Rectangle Steel Gage Block is a precision measuring tool designed for high accuracy applications. This 2 inch, AS-1 Grade block includes NIST certification, ensuring reliable and traceable measurements for quality control and calibration.
  • Rectangle shape
  • AS-1 accuracy grade
  • 2 inch size
  • Durable steel construction
  • NIST certified for traceability and reliability