Learn More
MSC Value Collection Gage Block, Rectangle, 6 in, Steel, Grade 0
Catalog No. 5000527212
Encompass
This steel gage block is designed for precision measurements, featuring a rectangular shape and 6-inch size with Grade 0 accuracy. It comes with NIST certification for reliable performance.
  • Rectangle shape for versatile use.
  • Grade 0 accuracy ensures high precision.
  • 6 inch size for standard measurement applications.
  • Constructed from durable steel.
  • Includes NIST certification for traceability.
Catalog No. 50-005-27212 Supplier MSC Supplier No. 86286200
This steel gage block is designed for precision measurements, featuring a rectangular shape and 6-inch size with Grade 0 accuracy. It comes with NIST certification for reliable performance.
  • Rectangle shape for versatile use.
  • Grade 0 accuracy ensures high precision.
  • 6 inch size for standard measurement applications.
  • Constructed from durable steel.
  • Includes NIST certification for traceability.