Learn More
MSC Value Collection Gage Block, Rectangle, Steel, 0.05 inch, Grade 0
Catalog No. 5000510878
Encompass
This steel gage block is designed for precision measurements, featuring a rectangular shape and a size of 0.05 " It meets Grade 0 accuracy standards and includes NIST certification, offering quality and value for various business needs.
  • Rectangle shape
  • Grade 0 accuracy
  • 0.05 inch size
  • Steel material
  • Includes NIST certification
  • Overall dimensions: 0.354 inch wide x 1.18 inch long
Catalog No. 50-005-10878 Supplier MSC Supplier No. 86280104
This steel gage block is designed for precision measurements, featuring a rectangular shape and a size of 0.05 " It meets Grade 0 accuracy standards and includes NIST certification, offering quality and value for various business needs.
  • Rectangle shape
  • Grade 0 accuracy
  • 0.05 inch size
  • Steel material
  • Includes NIST certification
  • Overall dimensions: 0.354 inch wide x 1.18 inch long