Learn More
MSC Value Collection Gage Block, Rectangle, Steel, 0.0508 Inch, Grade 0
Catalog No. 5000536819
Encompass
This is a high-precision rectangle steel gage block from Value Collection, ideal for calibration and inspection tasks requiring exceptional accuracy. It features a nominal size of 0.0508 inches and is manufactured to Grade 0 accuracy, ensuring reliability in critical measurements.
  • Rectangle shape for versatile use.
  • Constructed from durable steel material.
  • Precision measured at 0.0508 inches.
  • Certified to grade 0 accuracy.
  • Suitable for calibration and inspection applications.
Catalog No. 50-005-36819 Supplier MSC Supplier No. 86280328
This is a high-precision rectangle steel gage block from Value Collection, ideal for calibration and inspection tasks requiring exceptional accuracy. It features a nominal size of 0.0508 inches and is manufactured to Grade 0 accuracy, ensuring reliability in critical measurements.
  • Rectangle shape for versatile use.
  • Constructed from durable steel material.
  • Precision measured at 0.0508 inches.
  • Certified to grade 0 accuracy.
  • Suitable for calibration and inspection applications.