Learn More
MSC Value Collection Gage Block, Rectangle, Steel, 0.126 in, Grade 0
Supplier:  MSC 86282308
Encompass
This gage block features a rectangular design and is manufactured from steel, ensuring durability and stability for precise measurement applications. With an accuracy grade of 0 and a size of 0.126 inch, it delivers reliable performance. The product also includes NIST certification, affirming its high standards for accuracy.
  • Rectangular shape for versatile use
  • Constructed from steel for durability
  • Features accuracy grade 0 for high precision
  • Measures 0.126 inch for specific calibration needs
  • Includes NIST certification to confirm accuracy standards
Catalog No. 50-005-30510