Learn More
MSC Value Collection Gage Block, Round, Steel, 0.125 inch, Grade B
Catalog No. 5000529064
Encompass
This is a round steel gage block designed for precision measurements and spacing applications. It features an accuracy grade of B and is part of a collection known for providing quality at a great value.
  • Round shape
  • Grade B accuracy
  • Functions as a spacer block
  • Constructed from steel
  • 0.125 inch size
Catalog No. 50-005-29064 Supplier MSC Supplier No. 06464283
This is a round steel gage block designed for precision measurements and spacing applications. It features an accuracy grade of B and is part of a collection known for providing quality at a great value.
  • Round shape
  • Grade B accuracy
  • Functions as a spacer block
  • Constructed from steel
  • 0.125 inch size