Learn More
MSC Value Collection Gage Block Set, 36 Pc, 0.05 to 1 in, 0.0001 in Tolerance
Catalog No. 5000512807
Encompass
This gage block set contains 36 round pieces with a range of 0.05 to 1 " Designed for precision measurements, each block features a tolerance of 0.0001 inch and the set comes complete with a fitted case for storage and protection.
  • Round shape
  • 36 pieces
  • 0.05 to 1 inch range
  • 0.0001 inch tolerance
  • Includes fitted case
  • Not individually serialized
  • Country of origin: China
Catalog No. 50-005-12807 Supplier MSC Supplier No. 06464051
This gage block set contains 36 round pieces with a range of 0.05 to 1 " Designed for precision measurements, each block features a tolerance of 0.0001 inch and the set comes complete with a fitted case for storage and protection.
  • Round shape
  • 36 pieces
  • 0.05 to 1 inch range
  • 0.0001 inch tolerance
  • Includes fitted case
  • Not individually serialized
  • Country of origin: China