Learn More
MSC Value Collection Gage Block, Square, 0.132 inch, Grade 0, NIST Certified
Catalog No. 5000515987
Encompass
This gage block is a square steel block, precisely measuring 0.132 inch, and features an accuracy grade of 0. It includes NIST Certification for assured quality and traceability.
  • Square shape
  • Accuracy grade 0
  • Made from steel
  • Measures 0.132 inch
  • Includes NIST certification
Catalog No. 50-005-15987 Supplier MSC Supplier No. 86272606
This gage block is a square steel block, precisely measuring 0.132 inch, and features an accuracy grade of 0. It includes NIST Certification for assured quality and traceability.
  • Square shape
  • Accuracy grade 0
  • Made from steel
  • Measures 0.132 inch
  • Includes NIST certification