Learn More
MSC Value Collection Gage Block, Square, 0.135 in, Grade 0, Steel
Catalog No. 5000532091
Encompass
This gage block is a precision measuring tool, designed for accuracy and durability. It features a square shape and is made from steel, with a size of 0.135 in and an accuracy grade of 0. It includes NIST certification for guaranteed performance.
  • Square shape
  • Grade 0 accuracy
  • 0.135 in size
  • Steel material
  • NIST certified
Catalog No. 50-005-32091 Supplier MSC Supplier No. 86272754
This gage block is a precision measuring tool, designed for accuracy and durability. It features a square shape and is made from steel, with a size of 0.135 in and an accuracy grade of 0. It includes NIST certification for guaranteed performance.
  • Square shape
  • Grade 0 accuracy
  • 0.135 in size
  • Steel material
  • NIST certified