Learn More
MSC Value Collection Gage Block, Square, Grade 0, Steel, 0.10005 in
Catalog No. 5000510787
Encompass
The Value Collection Square Steel Gage Block is designed for precision measurement applications, offering an accuracy grade of 0. This high-quality gage block ensures reliable and consistent performance, making it an essential tool for maintaining dimensional accuracy in various industrial and calibration settings.
  • Ensures high precision in measurement tasks.
  • Constructed from durable steel for extended life.
  • Features a square shape for versatile application.
  • Meets Grade 0 accuracy standards for critical use.
Catalog No. 50-005-10787 Supplier MSC Supplier No. 86270964
The Value Collection Square Steel Gage Block is designed for precision measurement applications, offering an accuracy grade of 0. This high-quality gage block ensures reliable and consistent performance, making it an essential tool for maintaining dimensional accuracy in various industrial and calibration settings.
  • Ensures high precision in measurement tasks.
  • Constructed from durable steel for extended life.
  • Features a square shape for versatile application.
  • Meets Grade 0 accuracy standards for critical use.