Learn More
MSC Value Collection Gage Block, Square, Steel, 0.1005 in, Grade 0
Catalog No. 5000527569
Encompass
This steel gage block is designed for precise measurements. It features a square shape and an accuracy grade of 0, making it suitable for demanding metrology applications. Each gage block includes NIST certification for verified accuracy.
  • Includes NIST certification.
  • Made from durable steel.
  • Features a square shape.
  • Maintains an accuracy grade of 0.
  • Precisely sized at 0.1005 decimal "
Catalog No. 50-005-27569 Supplier MSC Supplier No. 86271053
Add to Cart
Add to Cart
This steel gage block is designed for precise measurements. It features a square shape and an accuracy grade of 0, making it suitable for demanding metrology applications. Each gage block includes NIST certification for verified accuracy.
  • Includes NIST certification.
  • Made from durable steel.
  • Features a square shape.
  • Maintains an accuracy grade of 0.
  • Precisely sized at 0.1005 decimal "