Learn More
MSC Value Collection Gage Block, Square, Steel, 0.6 in, Grade 0
Catalog No. 5000527232
Encompass
This Square Steel Gage Block is designed for precision measurement tasks. It features a 0.6 inch size and an Accuracy Grade of 0, making it suitable for demanding calibration and inspection applications. Crafted from durable steel, this gage block ensures reliability and longevity, backed by NIST certification for guaranteed accuracy.
  • Square shape for versatile measurement applications.
  • Accuracy grade of 0 ensures high precision and reliability.
  • Measures 0.6 inch, ideal for specific calibration needs.
  • Constructed from durable steel for long-lasting performance.
  • Includes NIST certification, providing traceability and assurance of accuracy.
Catalog No. 50-005-27232 Supplier MSC Supplier No. 86274909
This Square Steel Gage Block is designed for precision measurement tasks. It features a 0.6 inch size and an Accuracy Grade of 0, making it suitable for demanding calibration and inspection applications. Crafted from durable steel, this gage block ensures reliability and longevity, backed by NIST certification for guaranteed accuracy.
  • Square shape for versatile measurement applications.
  • Accuracy grade of 0 ensures high precision and reliability.
  • Measures 0.6 inch, ideal for specific calibration needs.
  • Constructed from durable steel for long-lasting performance.
  • Includes NIST certification, providing traceability and assurance of accuracy.