Learn More
MSC Value Collection Gage Block, Steel, Rectangle, 10 in, Grade 0, NIST Certified
Catalog No. 5000527231
Encompass
This product is a 10-inch, Grade 0, rectangle steel gage block. It is designed for precise measurement and calibration in industrial applications. The gage block includes NIST certification for assurance of accuracy.
  • Rectangle shape for versatile use
  • Grade 0 accuracy ensures high precision
  • Made from durable steel material
  • Includes NIST certification for traceability and quality assurance
  • 10 in size suitable for various measurements
Catalog No. 50-005-27231 Supplier MSC Supplier No. 86286507
This product is a 10-inch, Grade 0, rectangle steel gage block. It is designed for precise measurement and calibration in industrial applications. The gage block includes NIST certification for assurance of accuracy.
  • Rectangle shape for versatile use
  • Grade 0 accuracy ensures high precision
  • Made from durable steel material
  • Includes NIST certification for traceability and quality assurance
  • 10 in size suitable for various measurements