Learn More
MSC Value Collection Guidance Pulley, 4-1/2 in OD, 3/8 in Wire Rope, Galvanized Iron
Catalog No. 5000531534
Encompass
This single open side mount guidance pulley is designed to support various business needs with its quality construction. It features a 4-1/2 inch outside diameter and is suitable for 3/8 inch diameter wire ropes. The pulley is made from iron with a galvanized finish, ensuring durability.
  • Designed for wire rope applications.
  • Features a 4-1/2 inch outside diameter.
  • Accommodates 3/8 inch diameter wire ropes.
  • Constructed from durable iron.
  • Galvanized finish provides enhanced protection.
  • Mounting hole diameter ranges from 0.235 inch to 0.246 "
  • Manufactured in China.
Catalog No. 50-005-31534 Supplier MSC Supplier No. 67327981
This single open side mount guidance pulley is designed to support various business needs with its quality construction. It features a 4-1/2 inch outside diameter and is suitable for 3/8 inch diameter wire ropes. The pulley is made from iron with a galvanized finish, ensuring durability.
  • Designed for wire rope applications.
  • Features a 4-1/2 inch outside diameter.
  • Accommodates 3/8 inch diameter wire ropes.
  • Constructed from durable iron.
  • Galvanized finish provides enhanced protection.
  • Mounting hole diameter ranges from 0.235 inch to 0.246 "
  • Manufactured in China.