Learn More
MSC Value Collection Impulse Sealer, 8 in, 6 mil, 1 ea
Catalog No. 5000781624
Encompass
This tabletop thermal impulse sealer with cutter is designed for efficient sealing applications in various settings. It provides precise control for creating secure seals on materials up to 6 mil thick.
  • Tabletop thermal sealer with integrated cutter
  • Maximum seal length of 8 in
  • Capable of sealing materials up to 6 mil thick
  • Operates at 260 W with 110 V power supply
Catalog No. 50-007-81624 Supplier MSC Supplier No. 76218981
This tabletop thermal impulse sealer with cutter is designed for efficient sealing applications in various settings. It provides precise control for creating secure seals on materials up to 6 mil thick.
  • Tabletop thermal sealer with integrated cutter
  • Maximum seal length of 8 in
  • Capable of sealing materials up to 6 mil thick
  • Operates at 260 W with 110 V power supply