Learn More
MSC Value Collection Inside Caliper, 12 Inch Leg, Spring Joint, Steel, Polished
Catalog No. 5000529507
Encompass
This product is an inside caliper designed for precise measurement applications. It features a durable steel construction with a polished finish and a spring joint for accurate readings. The 12 inch flat legs are equipped with a quick nut adjustment for ease of use.
  • Inside caliper for accurate internal measurements
  • 12 inch leg length
  • Spring joint for reliable operation
  • Durable steel material
  • Polished finish for corrosion resistance
  • Flat legs for specific measurement tasks
  • Quick nut adjustment for rapid adjustments
  • Maximum measurement of 12 inches (300 mm)
Catalog No. 50-005-29507 Supplier MSC Supplier No. 06487128
This product is an inside caliper designed for precise measurement applications. It features a durable steel construction with a polished finish and a spring joint for accurate readings. The 12 inch flat legs are equipped with a quick nut adjustment for ease of use.
  • Inside caliper for accurate internal measurements
  • 12 inch leg length
  • Spring joint for reliable operation
  • Durable steel material
  • Polished finish for corrosion resistance
  • Flat legs for specific measurement tasks
  • Quick nut adjustment for rapid adjustments
  • Maximum measurement of 12 inches (300 mm)