Learn More
MSC Value Collection Inside Caliper, Spring Joint, Flat Leg, Steel, Polished
Catalog No. 5000512892
Encompass
This inside caliper is designed for precise internal measurements. Featuring an 8-inch (200mm) maximum measurement capacity, it utilizes a spring joint for smooth operation and a flat leg design for stability. The steel construction with a polished finish ensures durability and accuracy, further enhanced by a quick nut adjustment for efficient setting.
  • Features a spring joint for smooth and accurate operation.
  • Designed for precise inside measurements.
  • Equipped with a flat leg for stability and ease of use.
  • Constructed from durable steel.
  • Boasts a polished finish for enhanced appearance and corrosion resistance.
  • Includes quick nut adjustment for efficient and precise setting.
  • Offers an 8 inch (200 mm) maximum measurement capacity.
Catalog No. 50-005-12892 Supplier MSC Supplier No. 06487086
This inside caliper is designed for precise internal measurements. Featuring an 8-inch (200mm) maximum measurement capacity, it utilizes a spring joint for smooth operation and a flat leg design for stability. The steel construction with a polished finish ensures durability and accuracy, further enhanced by a quick nut adjustment for efficient setting.
  • Features a spring joint for smooth and accurate operation.
  • Designed for precise inside measurements.
  • Equipped with a flat leg for stability and ease of use.
  • Constructed from durable steel.
  • Boasts a polished finish for enhanced appearance and corrosion resistance.
  • Includes quick nut adjustment for efficient and precise setting.
  • Offers an 8 inch (200 mm) maximum measurement capacity.