Learn More
MSC Value Collection Inspection Surface Plate, 18 in x 24 in, 3 in Thick, Granite, Grade B
Catalog No. 5000514859
Encompass
This inspection surface plate is designed for precise measurements and inspections in various industrial settings. Crafted from high-quality granite, it provides a stable and accurate reference plane for quality control and manufacturing processes.
  • Dimensions: 18 in wide, 24 in long, 3 in thick
  • Constructed from granite for stability and accuracy
  • Grade B classification ensures reliable performance
  • Features a no ledge design
  • Overall unilateral flatness tolerance of 0.000150 in
  • Includes a certificate for quality assurance
  • Weighs 180 lb
  • Black color
  • Country of origin: China
Catalog No. 50-005-14859 Supplier MSC Supplier No. 00150037
This inspection surface plate is designed for precise measurements and inspections in various industrial settings. Crafted from high-quality granite, it provides a stable and accurate reference plane for quality control and manufacturing processes.
  • Dimensions: 18 in wide, 24 in long, 3 in thick
  • Constructed from granite for stability and accuracy
  • Grade B classification ensures reliable performance
  • Features a no ledge design
  • Overall unilateral flatness tolerance of 0.000150 in
  • Includes a certificate for quality assurance
  • Weighs 180 lb
  • Black color
  • Country of origin: China