Learn More
MSC Value Collection Inspection Surface Plate, 36 in Wide, 48 in Long, 4 in Thick, Granite, Grade B
Catalog No. 5000529747
Encompass
This inspection surface plate is crafted from durable granite, offering a reliable and precise surface for measurement and inspection tasks. With a grade B classification and an overall unilateral flatness tolerance of 0.000400 inch, it ensures high accuracy for critical applications. The plate measures 36 inches wide, 48 inches long, and 4 inches thick.
  • Includes a certificate of inspection
  • Black color
  • Constructed from granite material
  • Four inch thickness
  • Forty-eight inch overall length
  • Unilateral flatness tolerance of 0.000400 inch
  • Grade B classification
  • No ledge design
Catalog No. 50-005-29747 Supplier MSC Supplier No. 00150078
This inspection surface plate is crafted from durable granite, offering a reliable and precise surface for measurement and inspection tasks. With a grade B classification and an overall unilateral flatness tolerance of 0.000400 inch, it ensures high accuracy for critical applications. The plate measures 36 inches wide, 48 inches long, and 4 inches thick.
  • Includes a certificate of inspection
  • Black color
  • Constructed from granite material
  • Four inch thickness
  • Forty-eight inch overall length
  • Unilateral flatness tolerance of 0.000400 inch
  • Grade B classification
  • No ledge design