Learn More
MSC Value Collection Inspection Surface Plate, Granite, 2-Ledge, Grade B, 24 x 36 x 4 in
Catalog No. 5000530010
Encompass
This inspection surface plate is crafted from durable black granite, providing a stable and precise surface for measurement and inspection tasks. It features a 2-ledge design and meets Grade B flatness standards, ensuring accuracy for professional applications.
  • Crafted from durable black granite
  • 2-ledge design for versatility
  • Grade B flatness for reliable measurements
  • Overall dimensions: 24 in wide, 36 in long, 4 in thick
  • Unilateral flatness tolerance of 0.000100 in
  • Weighs 365 lb
  • Includes a certificate of inspection
Catalog No. 50-005-30010 Supplier MSC Supplier No. 00150060
This inspection surface plate is crafted from durable black granite, providing a stable and precise surface for measurement and inspection tasks. It features a 2-ledge design and meets Grade B flatness standards, ensuring accuracy for professional applications.
  • Crafted from durable black granite
  • 2-ledge design for versatility
  • Grade B flatness for reliable measurements
  • Overall dimensions: 24 in wide, 36 in long, 4 in thick
  • Unilateral flatness tolerance of 0.000100 in
  • Weighs 365 lb
  • Includes a certificate of inspection