Learn More
MSC Value Collection Inspection Surface Plate Stand, Stationary, Steel, 36 in Plate Length, 36 in High
Catalog No. 5000529582
Encompass
This is a stationary steel stand designed to support inspection surface plates. It features a robust construction and is ideal for maintaining the stability and precise positioning of surface plates in various industrial and laboratory settings.
  • Stationary stand type
  • Constructed from steel
  • Overall height of 36 inches
  • Designed for surface plates with a 36 inch length
  • Gray color finish
  • Fabricated from 2 x 2 x 1/4 angle iron
  • Compatible with MHC Surface Plates
Catalog No. 50-005-29582 Supplier MSC Supplier No. 00920553
This is a stationary steel stand designed to support inspection surface plates. It features a robust construction and is ideal for maintaining the stability and precise positioning of surface plates in various industrial and laboratory settings.
  • Stationary stand type
  • Constructed from steel
  • Overall height of 36 inches
  • Designed for surface plates with a 36 inch length
  • Gray color finish
  • Fabricated from 2 x 2 x 1/4 angle iron
  • Compatible with MHC Surface Plates