Learn More
MSC Value Collection Magnetic Indicator Base, 1-3/16 in Base Diameter, Holders Included
Catalog No. 5000530605
Encompass
This magnetic indicator base is designed to securely hold indicators and other instruments. It features a circular base with a diameter of 1-3/16 inches and provides 37 lbs of magnetic holding power on unpainted steel. Holders are included for immediate use.
  • Circular base type
  • 1-3/16 in base diameter
  • 37 lbs magnetic power on unpainted steel
  • Holders included
  • Accommodates holding an indicator in various ways
  • Compatible with other accessories for holding different instruments
Catalog No. 50-005-30605 Supplier MSC Supplier No. 63326474
This magnetic indicator base is designed to securely hold indicators and other instruments. It features a circular base with a diameter of 1-3/16 inches and provides 37 lbs of magnetic holding power on unpainted steel. Holders are included for immediate use.
  • Circular base type
  • 1-3/16 in base diameter
  • 37 lbs magnetic power on unpainted steel
  • Holders included
  • Accommodates holding an indicator in various ways
  • Compatible with other accessories for holding different instruments