Learn More
MSC Value Collection Measuring Wire, Thread Pitch, 48-3 TPI, 3 in Length
Catalog No. 5000529348
Encompass
This thread pitch diameter measuring wire from Value Collection is designed for precise measurement applications. It offers a wide range of thread pitch compatibility for both imperial and metric systems.
  • Accurate measurement of thread pitch diameter.
  • Compatible with thread pitches from 48 to 3 TPI.
  • Supports metric pitches from 0.5 to 6 mm.
  • Features a 3 in wire length for versatile use.
  • Supplied as a 48-piece set for comprehensive needs.
Catalog No. 50-005-29348 Supplier MSC Supplier No. 09182601
This thread pitch diameter measuring wire from Value Collection is designed for precise measurement applications. It offers a wide range of thread pitch compatibility for both imperial and metric systems.
  • Accurate measurement of thread pitch diameter.
  • Compatible with thread pitches from 48 to 3 TPI.
  • Supports metric pitches from 0.5 to 6 mm.
  • Features a 3 in wire length for versatile use.
  • Supplied as a 48-piece set for comprehensive needs.