Learn More
MSC Value Collection Motor Capacitor, 10 Microfarad, 440 Volt, 1 ea
Catalog No. 5000505579
Encompass
This motor capacitor provides reliable performance for various electrical applications. Designed for compatibility with 440 Volt systems, it offers a capacitance of 10 microfarads.
  • Compatible with 440 V systems
  • Features 10 microfarad capacitance
  • Dimensions: 2.16 in wide x 1.31 in deep x 3.75 in high
Catalog No. 50-005-05579 Supplier MSC Supplier No. 08687279
This motor capacitor provides reliable performance for various electrical applications. Designed for compatibility with 440 Volt systems, it offers a capacitance of 10 microfarads.
  • Compatible with 440 V systems
  • Features 10 microfarad capacitance
  • Dimensions: 2.16 in wide x 1.31 in deep x 3.75 in high