Learn More
MSC Value Collection Motor Capacitor, 5 Microfarad, 440 Volts, 1 ea
Catalog No. 5000504283
Encompass
This motor capacitor from Value Collection is designed to provide reliable performance in applications requiring a 5 microfarad capacitance at 440 volts.
  • Capacitance: 5 microfarad
  • Voltage: 440 volts
  • Dimensions: 2.16 in wide x 1.31 in deep x 2.38 in high
Catalog No. 50-005-04283 Supplier MSC Supplier No. 08687246
This motor capacitor from Value Collection is designed to provide reliable performance in applications requiring a 5 microfarad capacitance at 440 volts.
  • Capacitance: 5 microfarad
  • Voltage: 440 volts
  • Dimensions: 2.16 in wide x 1.31 in deep x 2.38 in high