Learn More
MSC Value Collection Motor Capacitor, 55 Microfarad, 440 Volts, 1 ea
Catalog No. 5000505781
Encompass
The motor capacitor is designed to optimize the performance of various motor applications. It offers a robust 440 volts capacity and a precise 55 microfarad capacitance, ensuring reliable and consistent operation. Its compact dimensions make it suitable for a range of installations.
  • Optimizes performance of various motor applications
  • Robust 440 volts capacity
  • Precise 55 microfarad capacitance
  • Ensures reliable and consistent operation
  • Compact dimensions: 2.91 in wide x 1.91 in deep x 4-3/4 in high
Catalog No. 50-005-05781 Supplier MSC Supplier No. 08687386
The motor capacitor is designed to optimize the performance of various motor applications. It offers a robust 440 volts capacity and a precise 55 microfarad capacitance, ensuring reliable and consistent operation. Its compact dimensions make it suitable for a range of installations.
  • Optimizes performance of various motor applications
  • Robust 440 volts capacity
  • Precise 55 microfarad capacitance
  • Ensures reliable and consistent operation
  • Compact dimensions: 2.91 in wide x 1.91 in deep x 4-3/4 in high