Learn More
MSC Value Collection Nitrile Exam Gloves, Medium, 3 mil, 1000/Pk.
Catalog No. 5000788406
Encompass
These disposable gloves are crafted from durable nitrile with a 3.0 mil thickness, suitable for medical applications. They are powder-free, non-sterile, and feature a smooth grip for enhanced tactile sensitivity.
  • Crafted from durable nitrile
  • Features a 3.0 mil thickness
  • Suitable for medical applications
  • Powder-free
  • Non-sterile
  • Smooth grip surface
  • Measures 9.5 in length
  • Medium glove size
  • White color
  • Ambidextrous design
  • Rolled cuff style
  • FDA approved
  • Manufactured in USA at an FDA registered factory
  • FDA 510K certified
Catalog No. 50-007-88406 Supplier MSC Supplier No. 10087724
These disposable gloves are crafted from durable nitrile with a 3.0 mil thickness, suitable for medical applications. They are powder-free, non-sterile, and feature a smooth grip for enhanced tactile sensitivity.
  • Crafted from durable nitrile
  • Features a 3.0 mil thickness
  • Suitable for medical applications
  • Powder-free
  • Non-sterile
  • Smooth grip surface
  • Measures 9.5 in length
  • Medium glove size
  • White color
  • Ambidextrous design
  • Rolled cuff style
  • FDA approved
  • Manufactured in USA at an FDA registered factory
  • FDA 510K certified