Learn More
MSC Value Collection Nylon Rope, Double Braid, 5/8 in, 1,275 Lb
Catalog No. 5000528223
Encompass
This durable rope is constructed from nylon with a double braid design, providing strength and reliability for various applications. It features a 5/8 in diameter and a 1,275 pound capacity. This item is sold in 1 ft increments and is available in total lengths that are multiples of 25 ft.
  • Constructed from nylon for durability
  • Double braid design
  • 5/8 in diameter
  • 1,275 Lb capacity
  • Priced in 1 ft increments
  • Available in lengths up to 600 ft, in multiples of 25 ft
  • Not returnable
  • Special handling and shipping dates apply for cut-to-length items
Catalog No. 50-005-28223 Supplier MSC Supplier No. 45901865
This durable rope is constructed from nylon with a double braid design, providing strength and reliability for various applications. It features a 5/8 in diameter and a 1,275 pound capacity. This item is sold in 1 ft increments and is available in total lengths that are multiples of 25 ft.
  • Constructed from nylon for durability
  • Double braid design
  • 5/8 in diameter
  • 1,275 Lb capacity
  • Priced in 1 ft increments
  • Available in lengths up to 600 ft, in multiples of 25 ft
  • Not returnable
  • Special handling and shipping dates apply for cut-to-length items