Learn More
MSC Value Collection Optical Comparator Chart & Reticle, 14 in, Radius/Angle, Mylar, 10x
Catalog No. 5000529177
Encompass
This optical comparator chart and reticle is designed for precise measurement applications with 10x magnification systems. Made from durable Mylar, it features a 14-inch diameter and is specifically configured for radius and angle measurements, providing clear and accurate readings for quality control and inspection tasks.
  • Designed for use with 10x magnification systems.
  • Enables accurate radius and angle measurements.
  • Features a 14 in overall diameter for comprehensive viewing.
  • Constructed from durable Mylar material.
Catalog No. 50-005-29177 Supplier MSC Supplier No. 04240255
This optical comparator chart and reticle is designed for precise measurement applications with 10x magnification systems. Made from durable Mylar, it features a 14-inch diameter and is specifically configured for radius and angle measurements, providing clear and accurate readings for quality control and inspection tasks.
  • Designed for use with 10x magnification systems.
  • Enables accurate radius and angle measurements.
  • Features a 14 in overall diameter for comprehensive viewing.
  • Constructed from durable Mylar material.