Learn More
MSC Value Collection Outside Caliper, 10 in, Flat Leg, Spring, Steel
Catalog No. 5000529543
Encompass
This outside caliper is constructed from steel with a polished finish and features a spring joint for accurate measurements. It provides a maximum measurement of 10 inches (250 mm) and includes a flat leg with quick nut adjustment. This tool is designed for rough measuring of diameters, especially on metal lathes, allowing users to efficiently gauge proximity to final dimensions.
  • Maximum measurement of 10 inches (250 mm)
  • Flat leg with quick nut adjustment for ease of use
  • Spring joint for consistent tension
  • Outside caliper type for external dimension measurement
  • Durable steel construction
  • Polished finish for longevity
  • Suitable for rough measuring diameters
  • Can measure up to 10-1/2 inches
Catalog No. 50-005-29543 Supplier MSC Supplier No. 06486104
This outside caliper is constructed from steel with a polished finish and features a spring joint for accurate measurements. It provides a maximum measurement of 10 inches (250 mm) and includes a flat leg with quick nut adjustment. This tool is designed for rough measuring of diameters, especially on metal lathes, allowing users to efficiently gauge proximity to final dimensions.
  • Maximum measurement of 10 inches (250 mm)
  • Flat leg with quick nut adjustment for ease of use
  • Spring joint for consistent tension
  • Outside caliper type for external dimension measurement
  • Durable steel construction
  • Polished finish for longevity
  • Suitable for rough measuring diameters
  • Can measure up to 10-1/2 inches