Learn More
MSC Value Collection Plug Gage Handle Bushing, 0.145-0.152 in
Catalog No. 5000535378
Encompass
This plug gage handle bushing offers reliable performance for your business needs. It is designed to hold plug gages with a measurement range of 0.145 to 0.152 inches, featuring precise outside diameters for secure fitting.
  • Minimum measurement of 0.145 in.
  • Maximum measurement of 0.152 in.
  • Large end outside diameter measures 0.25 in.
  • Small end outside diameter measures 0.20 in.
Catalog No. 50-005-35378 Supplier MSC Supplier No. 88800206
This plug gage handle bushing offers reliable performance for your business needs. It is designed to hold plug gages with a measurement range of 0.145 to 0.152 inches, featuring precise outside diameters for secure fitting.
  • Minimum measurement of 0.145 in.
  • Maximum measurement of 0.152 in.
  • Large end outside diameter measures 0.25 in.
  • Small end outside diameter measures 0.20 in.