Learn More
MSC Value Collection Plug Gage Handle, Single End, 0.075-0.18 in
Catalog No. 5000530208
Encompass
This single end plug gage handle is designed for precision measurement applications. It measures 2.15 inches long and 0.37 inches wide, and is intended for use with bushings.
  • Single end design for precise gauging
  • Measures a minimum of 0.075 inch
  • Measures a maximum of 0.180 inch
  • Requires bushings for proper operation
  • Manufactured in the United States
Catalog No. 50-005-30208 Supplier MSC Supplier No. 84745546
This single end plug gage handle is designed for precision measurement applications. It measures 2.15 inches long and 0.37 inches wide, and is intended for use with bushings.
  • Single end design for precise gauging
  • Measures a minimum of 0.075 inch
  • Measures a maximum of 0.180 inch
  • Requires bushings for proper operation
  • Manufactured in the United States